missing translation for 'onlineSavingsMsg'
Learn More

Anthralin, TRC

Product Code. 30333995
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30333995 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30333995 Supplier Toronto Research Chemicals Supplier No. TRCA6792001G
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dithranol
CAS 1143-38-0
Molecular Formula C14 H10 O3
Quantity 1 g
Synonym 1,8-Dihydroxy-9(10H)-anthracenone, 1,8-Dihydroxyanthrone, 1,8-Dihydroxy-9-anthrone, 1,8-Dihydroxyanthrone, Anthra-Derm, Anthralin, Batidrol, Batridol, Chrysodermol, Cignolin, Cigthranol, Derobin, Dithranol, DrithoCreme, Drithoscalp, Lasan, NSC 43970, NSC 629313, Psodadrate, Psoriacid-Stift, Psoriacide
InChI Formula InChI=1S/C14H10O3/c15-10-5-1-3-8-7-9-4-2-6-11(16)13(9)14(17)12(8)10/h1-6,15-16H,7H2
SMILES Oc1cccc2Cc3cccc(O)c3C(=O)c12
IUPAC Name 1,8-dihydroxy-10H-anthracen-9-one
Molecular Weight (g/mol) 226.23
Formula Weight 226.063
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.