missing translation for 'onlineSavingsMsg'
Learn More

Anthracene, TRC

Product Code. 30004771
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30004771 25 g 25g
17935095 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30004771 Supplier Toronto Research Chemicals Supplier No. TRCA67840025G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-12-7
Molecular Formula C14 H10
Molecular Weight (g/mol) 178.23
Synonym Anthracin, Green Oil, NSC 7958, Paranaphthalene, Tetra Olive N2G
IUPAC Name anthracene
SMILES c1ccc2cc3ccccc3cc2c1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C14H10/c1-2-6-12-10-14-8-4-3-7-13(14)9-11(12)5-1/h1-10H
Formula Weight 178.0783
Chemical Name or Material Anthracene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.