missing translation for 'onlineSavingsMsg'
Learn More

Anthracene, 99%, Thermo Scientific Chemicals

Catalog Number 11439522
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
120-12-7
C14H10
178.23
MFCD00001240
MWPLVEDNUUSJAV-UHFFFAOYSA-N
paranaphthalene, anthracin, green oil, anthracen, tetra olive n2g, anthracen german, anthrazen, anthracene, pure, bis-alkylamino, ccris 767
8418
CHEBI:35298
anthracene
C1=CC2=CC3=CC=CC=C3C=C2C=C1
This item is not returnable. View return policy
120-12-7
C14H10
178.23
MFCD00001240
MWPLVEDNUUSJAV-UHFFFAOYSA-N
paranaphthalene, anthracin, green oil, anthracen, tetra olive n2g, anthracen german, anthrazen, anthracene, pure, bis-alkylamino, ccris 767
8418
CHEBI:35298
anthracene
C1=CC2=CC3=CC=CC=C3C=C2C=C1

Anthracene is used in the production of the red dyealizarin and other dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Anthracene is used in the production of the red dyealizarin and other dyes.

Solubility
Soluble in alcohol, methanol (1.8 g/100 g), hexane (0.37 g/100 g). Insoluble in water.

Notes
Store in cool, dry conditions in well sealed containers. keep container tightly closed. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 214°C to 218°C
Density 1.25
Boiling Point 340°C to 342°C
Flash Point 121°C (249°F)
Quantity 25 g
UN Number UN3077
Beilstein 1905429
Merck Index 14,682
Solubility Information Soluble in alcohol,methanol (1.8g/100 g),hexane (0.37g/100 g). Insoluble in water.
Formula Weight 178.23
Percent Purity 99%
Chemical Name or Material Anthracene
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'