missing translation for 'onlineSavingsMsg'
Learn More

Anthracene, 90+%, Thermo Scientific™

Catalog Number 11358347
Click to view available options
Quantity:
500 g
2 kg
This item is not returnable. View return policy
120-12-7
C14H10
178.23
MFCD00001240
MWPLVEDNUUSJAV-UHFFFAOYSA-N
paranaphthalene, anthracin, green oil, anthracen, tetra olive n2g, anthracen german, anthrazen, anthracene, pure, bis-alkylamino, ccris 767
8418
CHEBI:35298
anthracene
C1=CC2=CC3=CC=CC=C3C=C2C=C1
This item is not returnable. View return policy
120-12-7
C14H10
178.23
MFCD00001240
MWPLVEDNUUSJAV-UHFFFAOYSA-N
paranaphthalene, anthracin, green oil, anthracen, tetra olive n2g, anthracen german, anthrazen, anthracene, pure, bis-alkylamino, ccris 767
8418
CHEBI:35298
anthracene
C1=CC2=CC3=CC=CC=C3C=C2C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 210°C to 215°C
Density 1.250 g/mL
Boiling Point 340°C to 342°C
Flash Point 121°C (249°F)
Quantity 500 g
UN Number UN3077
Beilstein 1905429
Merck Index 14,682
Solubility Information Insoluble in water.
Formula Weight 178.23
Percent Purity ≥90%
Physical Form Crystalline
Chemical Name or Material Anthracene
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'