missing translation for 'onlineSavingsMsg'
Learn More

Antennapedia Control Antibody, mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18415165 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18415165 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18415165 Supplier Novus Biologicals Supplier No. NBP223629MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antennapedia Control Polyclonal antibody specifically detects Antennapedia Control in Mouse, All samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Antennapedia Control
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias AP homeodomain, AP homeodomain Drosophilia
Host Species Rabbit
Immunogen Reacts with the Antennapedia homeodomain sequence RQIKIWFQNRRMKWKK, which is 100 % conserved across all known species.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse, All species
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.