missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Antennapedia Control Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-23629CL1
This item is not returnable.
View return policy
Description
Antennapedia Control Polyclonal antibody specifically detects Antennapedia Control in All,Mouse samples. It is validated for Western Blot
Specifications
| Antennapedia Control | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| All species, Mouse | |
| Purified |
| Western Blot | |
| CoraFluor 1 | |
| AP homeodomain, AP homeodomain Drosophilia | |
| Reacts with the Antennapedia homeodomain sequence RQIKIWFQNRRMKWKK, which is 100 % conserved across all known species. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction