missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Antalarmin hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Non-peptide corticotropin-releasing hormone receptor 1 (CRF1) antagonist (Ki = 1 nM). Suppresses CRF-induced ACTH secretion. Blocks CRF- and novelty-induced anxiety-like behavior in animal models of anxiety.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 415.01 |
| Purity | >99% |
| Molecular Weight (g/mol) | 415.022 |
| Chemical Name or Material | Antalarmin hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction