missing translation for 'onlineSavingsMsg'
Learn More

ANT Monoclonal Antibody (5F51BB5AG7), Agarose
GREENER_CHOICE

Product Code. 10450204
Change view
Click to view available options
Quantity:
250 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10450204 250 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10450204 Supplier Invitrogen Supplier No. 457525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

ANT Monoclonal Antibody (5F51BB5AG7), Agarose

ANT Monoclonal Antibody for IP

TRUSTED_SUSTAINABILITY

Specifications

Antigen ANT
Applications Immunoprecipitation
Classification Monoclonal
Clone 5F51BB5AG7
Conjugate Agarose
Formulation HEPES buffered saline with 0.02% sodium azide
Gene SLC25A4
Gene Accession No. P02722, P12235
Gene Alias 30 kDa calsequestrin-binding protein; 30 kDa CSQ-binding protein; AAC1; adenine nucleotide translocase-1; adenine nucleotide translocator 1; adenine nucleotide translocator 1 (skeletal muscle); adenine nucleotide translocator 1, skeletal muscle; ADP ATP translocase; ADP,ATP carrier protein 1; ADP,ATP carrier protein, heart; ADP,ATP carrier protein, heart isoform T1; ADP,ATP carrier protein, heart/skeletal muscle; ADP,ATP carrier protein, heart/skeletal muscle isoform T1; ADP/ATP carrier; ADP/ATP translocase; ADP/ATP translocase 1; ADP/ATP translocase T1; ADP/ATP-carrier protein; Anc1; ANT; ANT 1; ANT1; AU019225; CSQ-binding 30 kDa protein; fa22e07; heart/skeletal muscle ATP/ADP translocator; mANC1; mitochondrial adenine nucleotide translocator; MTDPS12; PEO2; PEO3; PEOA2; Slc25a4; SLC25A5; solute carrier family 25 (mitochondrial adenine nucleotide translocator) member 4; solute carrier family 25 (mitochondrial carrier, adenine nucleotide translocator), member 4; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 4; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 5; solute carrier family 25 member 4; T1; translocase; Unknown (protein for MGC:77591); wu:fa22e07; zgc:77591
Gene Symbols SLC25A4
Host Species Mouse
Immunogen Recombinant Human ANT.
Purification Method Purified
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 282478, 291
Target Species Human, Bovine
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Slurry
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.