missing translation for 'onlineSavingsMsg'
Learn More

Ansofaxine hydrochloride, MedChemExpress

Product Code. 16480159
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
20 mg
Unit Size:
10mg
20mg
5mg
This item is not returnable. View return policy

Product Code. 16480159

Brand: Medchem Express HYU00096

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ansofaxine hydrochloride (LY03005; LPM570065) is a triple reuptake inhibitor; inhibits serotonin, dopamine and norepinephrine reuptake with IC50 values of 723, 491 and 763 nM, respectively.

Ansofaxine hydrochloride (LY03005

Specifications

Chemical Name or Material Ansofaxine hydrochloride
CAS 916918-84-8
Color White
Purity Grade Notes Research
Molecular Formula C24H32ClNO3
Quantity 10 mg
Synonym LY03005 LPM570065
Solubility Information DMSO : 31.25 mg/mL (74.77 mM; Need ultrasonic) ∣H2O : 6.67 mg/mL (15.96 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(OC1=CC=C(C(C2(O)CCCCC2)CN(C)C)C=C1)C3=CC=C(C)C=C3.[H]Cl
Molecular Weight (g/mol) 417.97
Formula Weight 417.97
Percent Purity 99.87%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.