missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Anserine

Product Code. 16494768
Encompass_Preferred
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
missing translation for 'unitSize'
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494768 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 16494768 missing translation for 'mfr' MedChemExpress missing translation for 'supplierNo' HY113354

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Anserine, a methylated form of Carnosine, is an orally active, natural Histidine-containing dipeptide found in skeletal muscle of vertebrates. Anserine is not cleaved by serum carnosinase and act as biochemical buffers, chelators, antioxidants, and anti-glycation agents. Anserine improves memory functions in Alzheimer's disease (AD)-model mice.

Specifications

Purity 97.0%
CAS 584-85-0
Content And Storage 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
For Use With (Application) Metabolism-protein/nucleotide metabolism
Molecular Weight (g/mol) 240.26 g/mol
Product Type Natural Products
Quantity 10 mg
Shipping Condition Room temperature
Solubility H2O : 100 mg/mL (416.22 mM∣Need ultrasonic) ∣DMSO : <1 mg/mL (ultrasonic) (insoluble or slightly soluble)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.