missing translation for 'onlineSavingsMsg'
Learn More

Ansell ActivArmr™ 80-813 Foam-Coated Flame Resistant Gloves

Product Code. 12842253 Shop All Ansell Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12842253 11 12 pairs
12802253 6 12 pairs
12872243 7 12 pairs
12822253 8 12 pairs
12812253 9 12 pairs
12832253 10 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 12842253 Supplier Ansell Supplier No. 80813110
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Flame resistant gloves provide Arc Flash Hazard/Risk Category 2 protection against electrical arc flash hazards. Aramid Fiber shell allows for safe handling of sharp objects.

  • Aramid Fiber shell is inherently flame resistant, and allows for safe handling of sharp objects
  • Neoprene foam coating enhances grip in dry, wet, or oily conditions
  • Arc rating of 12 cal/cm2 meets NFPA 70E Hazard Risk Category 2
  • Ergonomic design is comfortable and flexible
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Arc Flash Level 2
Certifications/Compliance NFPA 70E Hazard Risk Category 2
Certifications/Compliance (Europe) EN 407: 412110
Construction Knit
Coating Material Neoprene
Color Green, Black
Cut Resistance Rating ANSI A4
Hand Specific
Material Aramid Fiber
Product Type Arc Flash Gloves
Size 11
EN388 Standard 2X42C
For Use With (Application) Automotive Assembly, Equipment/Machinery Operation, Mining, and Utility Work
Glove Cuff Style Knit
Puncture Rating ANSI/ISEA 105-16 Level 2
Series ActivArmr™ 80-813
Thumb Style Wing
Yarn Gauge 13
Length (English) 8.86 to 10.24 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.