missing translation for 'onlineSavingsMsg'
Learn More

Ansell ActivArmr™ 47-402 Nitrile Coated Medium Duty Gloves

Product Code. 12600912 Shop All Ansell Products
Click to view available options
Size:
7
8
9
10
Unit Size:
12 pairs
This item is not returnable. View return policy

Product Code. 12600912

Brand: Ansell 47402080

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Nitrile coating is abrasion and oil-resistant to enhance grip capabilities in dry or oily conditions. Gloves feature interlock knit lining, generous sizing, and a contoured design for increased comfort.

  • ActivArmr™ gloves combine the comfort of cotton gloves with the abrasion resistance and oil repellence of a nitrile-coated glove
  • Interlock knit lining, generous sizing, and contoured designs increase comfort
  • Special nitrile formulation provides mechanical resistance and oil repellency
  • Designed to conform to the hand's natural shape for long wear periods
  • Ensure cleanliness of metal sheets in the steel and motor industries
  • Ideal for a variety of medium-duty job tasks
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Coated Area Full Glove
Construction Cut and Sewn
Coating Material Nitrile
Color Blue, Natural
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material Cotton
Product Type Dip-coated Gloves
Size 8
For Use With (Application) Automotive Repairs, Heavy Equipment Operation, Metal Fabrication, and Chemical Handling
Glove Cuff Style Knit
Grip Design Smooth
Lining Lined
Packaging Type Polybag
Puncture Rating ANSI/ISEA 105-16 Level 2
Series 47-402
Thumb Style Wing
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.