missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-900 Nitrile Coated Oil Repellent Gloves

Product Code. 12690762
Click to view available options
Size:
6
7
8
9
10
This item is not returnable. View return policy
This item is not returnable. View return policy

Industrial work gloves designed for light mechanical protection, particularly in moderately oily manufacturing or maintenance environments.

  • Nitrile coating provides light mechanical protection and dexterity
  • Designed for use in moderately oily manufacturing and maintenance environments
  • Combines protection with enhanced grip and abrasion resistance
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category II
Coated Area Palm, Fingers
Coating Material Nitrile
Color White, Blue
Cut Resistance Rating ANSI A1
Exterior Finish No Texture
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size 8
For Use With (Application) Assembly, Equipment Operation, and Metal Fabrication
Glove Cuff Style Knit
Series 11-900
Yarn Gauge 13
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'