missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-900 Nitrile Coated Oil Repellent Gloves

Product Code. p-3348224 Shop All Ansell Products
Click to view available options
Size:
6
7
8
9
10
Unit Size:
12 pairs
This item is not returnable. View return policy

Product Code. 12670762

Brand: Ansell 11900060

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Industrial work gloves designed for light mechanical protection, particularly in moderately oily manufacturing or maintenance environments.

  • Nitrile coating provides light mechanical protection and dexterity
  • Designed for use in moderately oily manufacturing and maintenance environments
  • Combines protection with enhanced grip and abrasion resistance
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category II
Chlorinated No
Construction Knit
Cuff Material Nitrile
Coating Material Nitrile
Color White, Blue
Cut Resistance Rating ANSI A1
Exterior Finish No Texture
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size 6
For Use With (Application) Assembly, Equipment Operation, and Metal Fabrication
Gender Unisex
Glove Cuff Style Knit
Grip Design Rough
Lining Unlined
Series 11-900
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.