missing translation for 'onlineSavingsMsg'
Learn More

Ansamitocin P 3', MedChemExpress

Product Code. 16497938
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16497938

Brand: Medchem Express HY19839

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Ansamitocin P 3' exhibits antitumour activity, is an antibody drug conjugate cytotoxin.

Ansamitocin P 3' exhibits antitumour activity, is an antibody drug conjugate cytotoxin.

Specifications

Chemical Name or Material Ansamitocin P 3'
CAS 66547-09-9
Purity Grade Notes Research
Molecular Formula C32H43ClN2O9
Quantity 10 mg
Synonym Antibiotic C 15003P3' Maytansinol butyrate
SMILES C[C@]1([C@@](CC(N(C(C=C2C=C3OC)=C3Cl)C)=O)([H])OC(CCC)=O)[C@H]([C@@H]([C@](OC4=O)([H])C[C@]([C@](/C=C/C=C(C)/C2)([H])OC)(N4)O)C)O1
Molecular Weight (g/mol) 635.14
Formula Weight 635.14
Grade Research
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.