Learn More
Invitrogen™ ANR52 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121191
Description
Positive test controls include: U-251MG, HeLa, A-549, K-562, BT-474, Mouse brain, Mouse liver, Rat brain, Rat liver. Immunogen sequence: RRRDKFGRTP LHYAAANGSY QCAVTLVTAG AGVNEADCKG CSPLHYAAAS DTYRRAEPHT PSSHDAEEDE PLKESRRKEA FFCLEFLLDN GADPSLRDRQ GYTAVHYAAA YGNRQNLELL LEMSFNCLED VESTIPVSPL HLAAYNGHCE ALKTLAETLV NLDVRDHKGR TALFLATERG STECVEVLTA HGASALIKER KRKWTPLHAA A.
Putative regulatory subunit of protein phosphatase 6 (PP6) that may be involved in the recognition of phosphoprotein substrates. [UniProt].
Specifications
| ANR52 | |
| Polyclonal | |
| Unconjugated | |
| ANKRD52 | |
| 6430544C07; ANKRD33; ANKRD52; ankyrin repeat domain 33; ankyrin repeat domain 52; ankyrin repeat domain-containing protein 52; CVWG5837; G431002C21Rik; PP6-ARS-C; protein phosphatase 6 ankyrin repeat subunit C; RGD1307124; Serine/threonine-protein phosphatase 6 regulatory ankyrin repeat subunit C; serine/threonine-protein phosphatase 6 regulatory subunit ARS-C | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 237615, 283373, 362811 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.36 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8BTI7, Q8NB46 | |
| ANKRD52 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 450-660 of human ANKRD52 (NP_775866.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.