missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ANP32D Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575586
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
ANP32D (acidic (leucine-rich) nuclear phosphoprotein 32 family, member D), also known as PP32R2 (phosphoprotein 32-related protein 2) or tumorigenic protein pp32r2, is a 131 amino acid protein that contains 3 leucine-rich repeats and belongs to the ANP32 family. ANP32D shares 89.3% amino acid sequence identity with ANP32A, a tumor suppressor that inhibits a variety of cancers, such as prostate and breast cancers. Although similar to ANP32A, ANP32D is tumorigenic and is preferentially expressed in prostatic cancer. Conversely, it is uncertain that ANP32D is neurally expressed. Intronless, ANP32D may be a pseudogene that is expressed in pathological situations and may also modulate the oncogenic potential of human prostate cancer.
Specifications
| ANP32D | |
| Polyclonal | |
| Unconjugated | |
| ANP32D | |
| acidic (leucine-rich) nuclear phosphoprotein 32 family, member D; acidic leucine-rich nuclear phosphoprotein 32 family member D; acidic nuclear phosphoprotein 32 family member D; acidic nuclear phosphoprotein 32D; ANP32D; Phosphoprotein 32-related protein 2; pp32 related 2; PP32R2; Tumorigenic protein pp32r2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23519 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95626 | |
| ANP32D | |
| Synthetic peptide corresponding to residues in Human ANP32D. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction