missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ANP32B Recombinant Rabbit Monoclonal Antibody (K01_1W69)
GREENER_CHOICE

Product Code. 30827712
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30827712 100 μL
30828215 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30827712 Supplier Invitrogen™ Supplier No. MA563402
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

ANP32B Recombinant Monoclonal Antibody for ICC/IF, IHC (P), Western Blot

Multifunctional protein working as a cell cycle progression factor as well as a cell survival factor. Required for the progression from the G1 to the S phase. Anti-apoptotic protein which functions as a caspase-3 inhibitor. Has no phosphatase 2A (PP2A) inhibitor activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANP32B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_1W69
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene ANP32B
Gene Accession No. Q92688, Q9EST6
Gene Alias 2410015B15Rik; acidic (leucine-rich) nuclear phosphoprotein 32 family, member B; acidic leucine-rich nuclear phosphoprotein 32 family member B; acidic nuclear phosphoprotein 32 family member B; acidic nuclear phosphoprotein 32 family, member B; acidic protein rich in leucines; Anp32b; APRIL; OTTHUMP00000063770; Pal31; PHAPI2; PHAPI2a; proliferation related acidic leucine rich protein PAL31; proliferation-related acidic leucine-rich protein PAL31; Putative HLA-DR-associated protein I-2; silver-stainable protein SSP29; SSP29; TNF superfamily member 13
Gene Symbols ANP32B
Host Species Rabbit
Immunogen A synthetic peptide of human PHAPI2.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10541, 170724
Target Species Human, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.