missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANP32B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10843-1-AP-150UL
This item is not returnable.
View return policy
Description
Multifunctional protein working as a cell cycle progression factor as well as a cell survival factor. Required for the progression from the G1 to the S phase. Anti-apoptotic protein which functions as a caspase-3 inhibitor. Has no phosphatase 2A (PP2A) inhibitor activity.Specifications
| ANP32B | |
| Polyclonal | |
| Unconjugated | |
| ANP32B | |
| ANP32B, APRIL, PHAPI2, Silver stainable protein SSP29, SSP29 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10541, 67628 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q92688, Q9EST5 | |
| ANP32B | |
| ANP32B Fusion Protein Ag1274 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction