Learn More
ANP32A Mouse anti-Human, CoraLite 488, Clone: 2D8C5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67687-100UL
Description
ANP32A, an acidic leucine-rich nuclear phosphoprotein, is a member of the inhibitor of the histone acetyltransferase (INHAT) complex and preferentially binds to unmodified histone H3 tails in vitro. ANP32A is expressed in normal tissues as well as in pancreatic, breast and prostate cancer and other malignant tumors. ANP32A plays an important role in cell proliferation, apoptosis, transcriptional regulation, signal transduction and other processes.
Specifications
| ANP32A | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Western Blot | |
| 2D8C5 | |
| CoraLite 488 | |
| 100 μL | |
| 8125 | |
| Liquid |