missing translation for 'onlineSavingsMsg'
Learn More

ANP32A Antibody, DyLight 550, Novus Biologicals™

Product Code. 18405566 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18405566 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18405566 Supplier Novus Biologicals Supplier No. NBP281794R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ANP32A Polyclonal antibody specifically detects ANP32A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANP32A
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias acidic (leucine-rich) nuclear phosphoprotein 32 family, member A, acidic leucine-rich nuclear phosphoprotein 32 family member A, Acidic nuclear phosphoprotein pp32, cerebellar leucine rich acidic nuclear protein, I1PP2A, inhibitor-1 of protein phosphatase-2A, LANPPutative HLA-DR-associated protein I, Leucine-rich acidic nuclear protein, MAPMC15orf1, Mapmodulin, PHAP1MGC150373, PHAPIMGC119787, Potent heat-stable protein phosphatase 2A inhibitor I1PP2A, PP32, putative human HLA class II associated protein I
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 14 amino acids near the carboxy terminus of human ANP32A . The immunogen is located within the last 50 amino acids of ANP32A. Amino Acid Squence: KRKREPEDEGEDDD
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 8125
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.