missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ANP Monoclonal Antibody (GT1068)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531578
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Predicted reactivity: Mouse (80%), Rat (80%), Dog (85%), Pig (87%), Rabbit (81%), Bovine (85%). Positive Control: mouse heart. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene belongs to the natriuretic peptide family. Natriuretic peptides are implicated in the control of extracellular fluid volume and electrolyte homeostasis. This protein is synthesized as a large precursor (containing a signal peptide), which is processed to release a peptide from the N-terminus with similarity to vasoactive peptide, cardiodilatin, and another peptide from the C-terminus with natriuretic-diuretic activity. Mutations in this gene have been associated with atrial fibrillation familial type 6.
Specifications
| ANP | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 20% glycerol and no preservative | |
| P01160, P05125 | |
| NPPA | |
| Full length human ANP recombinant protein. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| GT1068 | |
| Unconjugated | |
| NPPA | |
| Anf; ANP; ATFB6; atrial natriuretic factor; Atrial natriuretic peptide; atriopeptin; Atriopeptin I; Atriopeptin II; Atriopeptin III; Atriopeptin-1; Atriopeptin-2; Atriopeptin-3; ATRST2; Auriculin-A; Auriculin-B; Cardiodilatin; Cardiodilatin-related peptide; cardionatrin; CDD; CDD-ANF; CDP; natriuretic peptide A; Natriuretic peptide precursor A (pronatriodilatin, also Anf, Pnd); natriuretic peptide precursor A variant 1; natriuretic peptide precursor A variant NPPA-M1; natriuretic peptide precursor A variant NPPA-M2; natriuretic peptide precursor A variant NPPA-M3; natriuretic peptide precursor type A; natriuretic peptide type A; natriuretic peptides A; Nppa; Pnd; Prepronatriodilatin; Pronatriodilatin; RATANF; urodilatin | |
| Mouse | |
| Protein G | |
| RUO | |
| 230899, 4878 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction