missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Anode Buffer Container Dx

Product Code. 15918826 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
4 Ea.
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15918826 4 Ea. Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15918826 Supplier Applied Biosystems™ Supplier No. 4393925
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Each package contains 4 buffer containers

The Anode Buffer Container Dx contains 1X running buffer to support electrophoresis applications on Applied Biosystems Dx genetic analyzers.

The Anode Buffer Container Dx is designed for use with Applied Biosystems Dx genetic analyzers.

The Anode Buffer Container Dx contains 1X running buffer to support all electrophoresis applications on Applied Biosystems Dx genetic analyzers. It is made in a ready-to-use, disposable container with a radio frequency identification (RFID) tag incorporated into the label. The top of the Anode Buffer Container Dx is heat-sealed with a plastic film, which should be removed prior to direct installation on to the instrument. Each package includes four individual containers.

This product is intended for in vitro diagnostic use on Applied Biosystems Dx genetic analyzers and is not validated for use with other systems or applications. Applied Biosystems Dx genetic analyzers are to be used by trained operators in the analysis of human DNA for the detection of fluorescently labeled DNA nucleotides by capillary electrophoresis. Refer to the appropriate Applied Biosystems Dx genetic analyzer user guide for further details.

This is an anode buffer container designed for use with the 3500 Dx/3500xL Dx Genetic Analyzer. The 3500 Dx series conditioning reagent, anode buffer, cathode buffer and septa cathode buffer are required for instrument operation.

Order Info

4 Pack

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Each package contains 4 buffer containers.
Store at 2 - 8°C.
For Use With (Equipment) 3500xL Dx Genetic Analyzer, 3500 Dx Genetic Analyzer
For Use With (Application) Sequencing
Product Type Anode Buffer Container
Quantity 4 Ea.
Shipping Condition Wet Ice

For In Vitro Diagnostic Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.