missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Anode Buffer Container (ABC) for 3500 Series Systems for Protein Quality Analysis

Product Code. 15879876 Shop All Applied Biosystems Products
Click to view available options
Quantity:
4 Containers
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15879876

Brand: Applied Biosystems™ A31278

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Contains 4 anode buffer containers.

The anode buffer container (ABC) contains 1X running buffer to support protein quality analysis applications on the Applied Biosystems™ 3500 series genetic analyzers for protein quality analysis.

  • Each package includes four containers
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains 4 Anode Buffer containers.
Store at 2–8°C until ready to use. Refer to the expiration date on the label.
For Use With (Equipment) 3500 Genetic Analyzer for Protein Quality Analysis, 3500xL Genetic Analyzer for Protein Quality Analysis
For Use With (Application) Sequencing
Product Type Anode Buffer Container
Quantity 4 Containers
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.