missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ANO7 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA557378

Product Code. 15785271

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: VRTGLYCRDQ AHAERWAMTS ETSSGSHCAR SRMLRRRAQE EDSTVLIDVS PPEAEKRGSY GSTAHASEPG GQQ Highest antigen sequence identity to the following orthologs: Mouse - 37%, Rat - 37%.

Has calcium-dependent phospholipid scramblase activity; scrambles phosphatidylserine, phosphatidylcholine and galactosylceramide. Does not exhibit calcium-activated chloride channel (CaCC) activity. May play a role in cell-cell interactions.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ANO7
Polyclonal
Unconjugated
ANO7
ANO7; anoctamin 7; anoctamin-7; Dresden transmembrane protein of the prostate; Dresden-transmembrane protein of the prostate; DTMPP; D-TMPP; IPCA5; IPCA-5; New gene expressed in prostate; NGEP; PCANAP5; PCANAP5L; prostate cancer associated protein 5; Prostate cancer-associated protein 5; TMEM16G; Transmembrane protein 16G
Rabbit
Antigen Affinity Chromatography
RUO
50636
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q6IWH7
ANO7
Recombinant protein corresponding to Human ANO7. Recombinant protein control fragment (Product #RP-96135).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.