missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ANO7 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15674945
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15674945 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15674945 Supplier Invitrogen™ Supplier No. PA551025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ANO7 Polyclonal Antibody detects endogenous levels of ANO7 protein.

Has calcium-dependent phospholipid scramblase activity; scrambles phosphatidylserine, phosphatidylcholine and galactosylceramide. Does not exhibit calcium-activated chloride channel (CaCC) activity. May play a role in cell-cell interactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANO7
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ANO7
Gene Accession No. Q6IWH7
Gene Alias ANO7; anoctamin 7; anoctamin-7; Dresden transmembrane protein of the prostate; Dresden-transmembrane protein of the prostate; DTMPP; D-TMPP; IPCA5; IPCA-5; New gene expressed in prostate; NGEP; PCANAP5; PCANAP5L; prostate cancer associated protein 5; Prostate cancer-associated protein 5; TMEM16G; Transmembrane protein 16G
Gene Symbols ANO7
Host Species Rabbit
Immunogen Synthesized peptide derived from the C-terminal region of human NGEP.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50636
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.