missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ANO7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549839
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total ANO7 protein.
Has calcium-dependent phospholipid scramblase activity; scrambles phosphatidylserine, phosphatidylcholine and galactosylceramide. Does not exhibit calcium-activated chloride channel (CaCC) activity. May play a role in cell-cell interactions.
Specifications
| ANO7 | |
| Polyclonal | |
| Unconjugated | |
| ANO7 | |
| ANO7; anoctamin 7; anoctamin-7; Dresden transmembrane protein of the prostate; Dresden-transmembrane protein of the prostate; DTMPP; D-TMPP; IPCA5; IPCA-5; New gene expressed in prostate; NGEP; PCANAP5; PCANAP5L; prostate cancer associated protein 5; Prostate cancer-associated protein 5; TMEM16G; Transmembrane protein 16G | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 50636 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6IWH7 | |
| ANO7 | |
| Synthesized peptide derived from C-terminal of human TM16G. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction