missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANO2 Antibody (CL10631) [Alexa Fluor™ 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21178AF350
This item is not returnable.
View return policy
Description
ANO2 Monoclonal antibody specifically detects ANO2 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| ANO2 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| anoctamin 2, anoctamin-2, DKFZp434P102, TMEM16B, transmembrane protein 16B (eight membrane-spanning domains) | |
| This antibody was developed using a recombinant protein derived from Q9NQ90, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Neuroscience | |
| 57101 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL10631 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction