missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Annexin V Recombinant Protein, FITC, eBioscience™

Product Code. 15541917
Change view
Click to view available options
Quantity:
100 Tests
20 Tests
300 Tests
Unit Size:
100 tests
20 tests
300 tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15541917 300 Tests 300 tests
15531917 20 Tests 20 tests
15521917 100 Tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15541917 Supplier Invitrogen™ Supplier No. BMS306FI300
  • ONLINE EXCLUSIVE PRICE
    £322.00
    £241.65 / 300 tests
    Save £80.35
    24% Off
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recombinant Protein

The protein encoded by this gene belongs to the annexin family of calcium-dependent phospholipid binding proteins some of which have been implicated in membrane-related events along exocytotic and endocytotic pathways. Annexin 5 is a phospholipase A2 and protein kinase C inhibitory protein with calcium channel activity and a potential role in cellular signal transduction, inflammation, growth and differentiation. Annexin 5 has also been described as placental anticoagulant protein I, vascular anticoagulant-alpha, endonexin II, lipocortin V, placental protein 4 and anchorin CII. The gene spans 29 kb containing 13 exons, and encodes a single transcript of approximately 1.6 kb and a protein product with a molecular weight of about 35 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 300 Tests
For Use With (Application) Flow Cytometry
Molecular Weight (g/mol) 35.8kDa
Source E. coli
Structural Form E. Coli
Biological Activity Annexin V exhibits anti-phospholipase activity and binds to phosphatidylserine (PS).
Specific Reactivity All
Cross Reactivity All
Species E. coli
Protein Subtype Recombinant
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.