missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin V Mouse anti-Human, Mouse, Clone: 1E6A8, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | Annexin V |
|---|---|
| Clone | 1E6A8 |
| Concentration | 0.93 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry, Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16827945
|
Proteintech
66245-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16817945
|
Proteintech
66245-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene belongs to the annexin family of calcium-dependent phospholipid binding proteins some of which have been implicated in membrane-related events along exocytotic and endocytotic pathways. Annexin 5 is a phospholipase A2 and protein kinase C inhibitory protein with calcium channel activity and a potential role in cellular signal transduction, inflammation, growth and differentiation. Annexin 5 has also been described as placental anticoagulant protein I, vascular anticoagulant-alpha, endonexin II, lipocortin V, placental protein 4 and anchorin CII. The gene spans 29 kb containing 13 exons, and encodes a single transcript of approximately 1.6 kb and a protein product with a molecular weight of about 35 kDa.Specifications
| Annexin V | |
| 0.93 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Anchorin CII, Annexin 5, annexin A5, Annexin V, ANX5, ANXA5, Calphobindin I, CBP I, Endonexin II, ENX2, Lipocortin V, PAP I, PP4, Thromboplastin inhibitor, VAC alpha, Vascular anticoagulant alpha | |
| Anxa5 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 1E6A8 | |
| Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry, Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Mouse, Human | |
| P08758, P48036 | |
| 11747, 308 | |
| Annexin V Fusion Protein Ag1538 | |
| Primary | |
| -20°C | |
| ANXA5 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title