missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Annexin V Monoclonal Antibody (C13)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA543958
This item is not returnable.
View return policy
Description
Annexin V Monoclonal Antibody for ICC/IF, IHC (P), Western Blot
The protein encoded by this gene belongs to the annexin family of calcium-dependent phospholipid binding proteins some of which have been implicated in membrane-related events along exocytotic and endocytotic pathways. Annexin 5 is a phospholipase A2 and protein kinase C inhibitory protein with calcium channel activity and a potential role in cellular signal transduction, inflammation, growth and differentiation. Annexin 5 has also been described as placental anticoagulant protein I, vascular anticoagulant-alpha, endonexin II, lipocortin V, placental protein 4 and anchorin CII. The gene spans 29 kb containing 13 exons, and encodes a single transcript of approximately 1.6 kb and a protein product with a molecular weight of about 35 kDa.
Specifications
| Annexin V | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P08758, P48036, P81287 | |
| Anxa5 | |
| Recombinant protein Annexin V. The antigen corresponds to amino acid range 1-320 of the target protein. | |
| 20 μL | |
| Primary | |
| Human, Mouse, Canine, Rabbit, Bovine, Sheep, Pig, Horse | |
| Antibody | |
| IgG1 κ |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| C13 | |
| Unconjugated | |
| Anxa5 | |
| Anchorin CII; annexin 5; annexin A5; Annexin V; annexin-5; Anx5; ANXA5; Calphobindin I; CBP-I; Endonexin II; ENX2; epididymis secretory protein Li 7; HEL-S-7; LC5; lipocortin V; PAP-I; placental anticoagulant protein 4; Placental anticoagulant protein I; PP4; R74653; RPRGL3; Thromboplastin inhibitor; VAC-alpha; Vascular anticoagulant-alpha | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 100072241, 100342904, 100521982, 101114445, 11747, 281626, 308, 476094 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction