missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin V Antibody (3B9F3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52413
This item is not returnable.
View return policy
Description
Annexin V Monoclonal specifically detects Annexin V in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
Specifications
| Annexin V | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready | |
| Anchorin CII, annexin A5, Annexin V, annexin-5, ANX5, Calphobindin I, Endonexin II, ENX2CBP-I, Lipocortin V, PAP-I, Placental anticoagulant protein 4, Placental anticoagulant protein I, PP4, Thromboplastin inhibitor, VAC-alpha, Vascular anticoagulant-alpha | |
| Mouse | |
| 36 kDa | |
| 0.1 mg | |
| Apoptosis, Apoptosis Detections | |
| 308 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry | |
| 3B9F3 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| ANXA5 | |
| Purified recombinant fragment of human ANXA5 (AA: 160-320) expressed in E. Coli. | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction