missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A9 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99556AF700
This item is not returnable.
View return policy
Description
Annexin A9 Polyclonal antibody specifically detects Annexin A9 in Human samples. It is validated for Western Blot
Specifications
| Annexin A9 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 8416 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 700 | |
| annexin 31, annexin A9, Annexin XXXI, Annexin-31, Annexin-9, ANX31pemphaxin, Pemphaxin | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Annexin A9. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction