missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A8/ANXA8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-57118
This item is not returnable.
View return policy
Description
Annexin A8/ANXA8 Polyclonal specifically detects Annexin A8/ANXA8 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Annexin A8/ANXA8 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
| annexin A8, Annexin VIII, Annexin-8, ANX8, FLJ53095, VAC-beta, Vascular anticoagulant-beta | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| ANXA8 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MAWWKSWIEQEGVTVKSSSHFNPDPDAETLYQAMK | |
| 100 μL | |
| Signal Transduction | |
| 653145 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction