missing translation for 'onlineSavingsMsg'
Learn More

Annexin A8/ANXA8 Antibody [DyLight 350], Novus Biologicals™

Product Code. 30736373 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30736373 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736373 Supplier Novus Biologicals Supplier No. AF8105D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Annexin A8/ANXA8 Polyclonal antibody specifically detects Annexin A8/ANXA8 in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Annexin A8/ANXA8
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias annexin A8, Annexin VIII, Annexin-8, ANX8, FLJ53095, VAC-beta, Vascular anticoagulant-beta
Host Species Sheep
Immunogen E. coli-derived recombinant human Annexin A8/ANXA8, Ala2-Pro327, Accession # P13928
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 653145
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.