missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A8/ANXA8 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05654-20ul
This item is not returnable.
View return policy
Description
Annexin A8/ANXA8 Polyclonal antibody specifically detects Annexin A8/ANXA8 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Annexin A8/ANXA8 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| annexin A8, Annexin VIII, Annexin-8, ANX8, FLJ53095, VAC-beta, Vascular anticoagulant-beta | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-276 of human ANXA8L2 (NP_001265853.1). MAWWKAWIEQEGVTVKSSSHFNPDPDAETLYKAMKGIGVGSQLLSHQAAAFAFPSSALTSVSPWGQQGHLCCNPAGTNEQAIIDVLTKRSNTQRQQIAKSFKAQFGKDLTETLKSELSGKFERLIVALMYPPYRYEAKELHDAMKGSRDDVSSFVDPALALQDAQDLYAAGEKIRGTDEMKFITILCTRSATHLLRVKCTQNLHSYFAERLYYAMKGAGTRDGTLIRNIVSRSEIDLNLIKCHFKKMYGKTLSSMIMEDTSGDYKNALLSLVGSDP | |
| 20 μg | |
| Signal Transduction | |
| 653145 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction