missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Annexin A6 Monoclonal Antibody (4G3)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA547335
This item is not returnable.
View return policy
Description
Annexin A6 Monoclonal Antibody for Western Blot, ICC/IF
Annexin VI belongs to a family of calcium-dependent membrane and phospholipid binding proteins. Although their functions are still not clearly defined, several members of the annexin family have been implicated in membrane-related events along exocytotic and endocytotic pathways. The annexin VI gene is approximately 60 kbp long and contains 26 exons. It encodes a protein of about 68 kDa that consists of eight 68-amino acid repeats separated by linking sequences of variable lengths. It is highly similar to human annexins I and II sequences, each of which contain four such repeats. Exon 21 of annexin VI is alternatively spliced, giving rise to two isoforms that differ by a 6-amino acid insertion at the start of the seventh repeat. Annexin VI has been implicated in mediating the endosome aggregation and vesicle fusion in secreting epithelia during exocytosis.
Specifications
| Annexin A6 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 5mM sodium azide | |
| P08133, P14824, P48037 | |
| ANXA6 | |
| Recombinant full length annexin A6 expressed in and purified from E. coli. | |
| 500 μL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey, Horse | |
| Antibody | |
| IgG1 |
| Western Blot, Immunocytochemistry | |
| 4G3 | |
| Unconjugated | |
| ANXA6 | |
| 67 kDa calelectrin; Annexin A6; annexin VI; annexin VI (p68); annexin VI, p68; annexin-6; Anx6; ANXA6; AnxVI; AW107198; Cabm; calcium-binding protein 65/67; calcium-binding protein CATA 65/67; calcium-binding protein p68; calelectrin; calphobindin II; calphobindin-II; Camb; CBP 65/67; CBP68; chromobindin-20; CPB-II; lipocortin VI; p68; p70; Protein III; testis secretory sperm-binding protein Li 198a | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 100071569, 11749, 309, 479325, 79125 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction