missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98751-50ul
This item is not returnable.
View return policy
Description
Annexin A6 Polyclonal antibody specifically detects Annexin A6 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
| Annexin A6 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry-Paraffin 1:2500-1:10000 | |
| 67 kDa calelectrin, annexin A6, Annexin VI, annexin VI (p68), annexin-6, ANX6, calcium-binding protein p68, calelectrin, calphobindin II, calphobindin-II, CBP68, Chromobindin-20, CPB-II, Lipocortin VI, p68, p70, Protein III | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the human Annexin A6. | |
| 50 μg | |
| Endosome Markers | |
| 309 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction