missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Annexin A4 Recombinant Rabbit Monoclonal Antibody (24GB4715)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557334
This item is not returnable.
View return policy
Description
Annexin A4 Recombinant Monoclonal Antibody for Western Blot, Flow
Annexin IV (ANX4) belongs to the annexin family of calcium-dependent phospholipid binding proteins. Although their functions are still not clearly defined, several members of the annexin family have been implicated in membrane-related events along exocytotic and endocytotic pathways. ANX4 has 45 to 59% identity with other members of its family and shares a similar size and exon-intron organization. Isolated from human placenta, ANX4 encodes a protein that has possible interactions with ATP, and has in vitro anticoagulant activity and also inhibits phospholipase A2 activity. ANX4 is almost exclusively expressed in epithelial cells.
Specifications
| Annexin A4 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| ANXA4 | |
| 35-beta calcimedin; 36 kDa zymogen granule membrane-associated protein; AI265406; AIV; annexin 4; annexin A4; Annexin IV; annexin IV (placental anticoagulant protein II); annexin-4; ANX IV; ANX4; ANXA 4; Anxa4; ANXIV; AW106930; Carbohydrate-binding protein p33/p41; chromobindin-4; endonexin; endonexin I; epididymis secretory protein Li 274; HEL-S-274; Lipocortin IV; P32.5; p33/41 (annexin IV); PAP-II; PIG28; Placental anticoagulant protein II; PP4-X; proliferation-inducing gene 28; proliferation-inducing protein 28; Protein II; unnamed protein product; Xanx-4; ZAP 36/annexin IV; ZAP36; zymogen granule membrane associated protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11746, 307, 79124 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB4715 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P09525, P55260, P97429 | |
| ANXA4 | |
| Recombinant protein of human Annexin-4/ANXA4 (aa 2-319). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction