missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Annexin A4 Monoclonal Antibody (C2), Invitrogen™

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA551045
This item is not returnable.
View return policy
Description
Annexin IV (ANX4) belongs to the annexin family of calcium-dependent phospholipid binding proteins. Although their functions are still not clearly defined, several members of the annexin family have been implicated in membrane-related events along exocytotic and endocytotic pathways. ANX4 has 45 to 59% identity with other members of its family and shares a similar size and exon-intron organization. Isolated from human placenta, ANX4 encodes a protein that has possible interactions with ATP, and has in vitro anticoagulant activity and also inhibits phospholipase A2 activity. ANX4 is almost exclusively expressed in epithelial cells.
Specifications
| Annexin A4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| P08132, P09525 | |
| ANXA4 | |
| Recombinant Human ANXA4 protein, Ala2-Asp319 (Accession #P09525). | |
| 200 μL | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG1 κ |
| Western Blot, Immunohistochemistry (Paraffin), Western Blot | |
| C2 | |
| Unconjugated | |
| ANXA4 | |
| 35-beta calcimedin; 36 kDa zymogen granule membrane-associated protein; AI265406; AIV; annexin 4; annexin A4; Annexin IV; annexin IV (placental anticoagulant protein II); annexin-4; ANX IV; ANX4; ANXA 4; Anxa4; ANXIV; AW106930; Carbohydrate-binding protein p33/p41; chromobindin-4; endonexin; endonexin I; epididymis secretory protein Li 274; HEL-S-274; Lipocortin IV; P32.5; p33/41 (annexin IV); PAP-II; PIG28; Placental anticoagulant protein II; PP4-X; proliferation-inducing gene 28; proliferation-inducing protein 28; Protein II; unnamed protein product; Xanx-4; ZAP 36/annexin IV; ZAP36; zymogen granule membrane associated protein | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 100312960, 307 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction