missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A4 Antibody (SAIC-14C-10F12) - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
£190.00 - £494.00
Specifications
| Antigen | Annexin A4 |
|---|---|
| Clone | SAIC-14C-10F12 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Mass Spectrometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18692225
|
Novus Biologicals
NBP3-20093-0.2mg |
0.2 mg |
£494.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18654944
|
Novus Biologicals
NBP3-20093-0.025mg |
0.025 mg |
£190.00
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Annexin A4 Monoclonal antibody specifically detects Annexin A4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Mass SpectrometrySpecifications
| Annexin A4 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| annexin A4, Annexin IV, annexin IV (placental anticoagulant protein II), annexin-4, ANX4Carbohydrate-binding protein p33/p41, Chromobindin-4, chromobindin-4,35-beta calcimedin, DKFZp686H02120, Endonexin I, Lipocortin IV, MGC75105, P32.5, PAP-II, PIG28, Placental anticoagulant protein II, PP4-X, proliferation-inducing gene 28, proliferation-inducing protein 28, Protein II, ZAP36 | |
| Peptide 'DEGNYLDDALVR' derived from Annexin A4 conjugated to KLH. | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
| SAIC-14C-10F12 | |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Mass Spectrometry | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| PBS | |
| 307 | |
| IgG κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title