missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Annexin A3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527884
This item is not returnable.
View return policy
Description
Recommended positive controls: A549, HCT116. Predicted reactivity: Mouse (91%), Rat (86%), Bovine (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. This protein functions in the inhibition of phopholipase A2 and cleavage of inositol 1,2-cyclic phosphate to form inositol 1-phosphate. This protein may also play a role in anti-coagulation.
Specifications
| Annexin A3 | |
| Polyclonal | |
| Unconjugated | |
| ANXA3 | |
| 35-alpha calcimedin; Annexin 3; annexin A3; Annexin III; Annexin III (Lipocortin III); annexin III (lipocortin III, 1,2-cyclic-inositol-phosphate phosphodiesterase, placental anticoagulant protein III, calcimedin 35-alpha); Annexin3; annexin-3; ANX3; ANXA 3; ANXA3; calcimedin 35-alpha; inositol 1,2-cyclic phosphate 2-phosphohydrolase; LC3; Lipocortin III; LRRGT00047; PAP-III; Placental anticoagulant protein III | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 25291, 306 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| P12429, P14669 | |
| ANXA3 | |
| Recombinant protein encompassing a sequence within the center region of human Annexin III. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction