missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Annexin A13 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553835
This item is not returnable.
View return policy
Description
Immunogen sequence: RHSQSYTLSE GSQQLPKGDS QPSTVVQPLS HPSRNGEPEA PQPAKASSPQ GFDVDRDAKK LNKACKGMGT N Highest antigen sequence identity to the following orthologs: Mouse - 32%, Rat - 32%.
This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. The specific function of this gene has not yet been determined; however, it is associated with the plasma membrane of undifferentiated, proliferating endothelial cells and differentiated villus enterocytes. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
| Annexin A13 | |
| Polyclonal | |
| Unconjugated | |
| ANXA13 | |
| 1810034H17Rik; Annexin A13; annexin XIII; annexin, intestine-specific; annexin-13; ANX13; Anxa13; AV055219; Intestine-specific annexin; ISA | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 312 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P27216 | |
| ANXA13 | |
| Recombinant protein corresponding to Human Annexin A13. Recombinant protein control fragment (Product #RP-92286). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ser du en mulighed for forbedring?Del en indholdskorrektion