missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10479-2-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the annexin family, a group of calcium-dependent phospholipid-binding proteins. Annexins have unique N-terminal domains and conserved C-terminal domains, which contain the calcium-dependent phospholipid-binding sites. The encoded protein is a 56-kD antigen recognized by sera from patients with various autoimmune diseases. Several transcript variants encoding two different isoforms have been identified.Specifications
| Annexin A11 | |
| Polyclonal | |
| Unconjugated | |
| ANXA11 | |
| 56 kDa autoantigen, Annexin 11, annexin A11, Annexin XI, ANX11, ANXA11, CAP 50, CAP50 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11744, 290527, 311 | |
| -20°C | |
| Liquid |
| Western Blot, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P50995, P97384 | |
| Anxa11 | |
| Annexin A11 Fusion Protein Ag0729 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction