missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Annexin A11 Protein

Product Code. 18282890 Shop All Bio Techne Products
Click to view available options
Quantity:
250 μg
Unit Size:
250µg
This item is not returnable. View return policy

Product Code. 18282890

Brand: Novus Biologicals™ NBP150857250UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-505 of Human Annexin A11 The Recombinant Human Annexin A11 Protein is derived from E. coli. The Recombinant Human Annexin A11 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.25 mg/mL
For Use With (Application) SDS-PAGE
Formulation PBS containing 20% glycerol, 2 mM DTT, 100 mM NaCl
Molecular Weight (g/mol) 56.6 kDa
Purification Method SDS-PAGE
Quantity 250 μg
Source E. coli
Immunogen ANXA11, 1-505 aa, Human, His tag, E. coli
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name Annexin A11
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.