missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Annexin A11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582983
This item is not returnable.
View return policy
Description
Immunogen sequence: QPPVTYPGQP PVPLPGQQQP VPSYPGYPGS GTVTPAVPPT QFGSRGTITD APGFDPLRDA EVLRKAMKGF GTDEQ Highest antigen sequence indentity to the following orthologs: Mouse - 84%, Rat - 85%.
This gene encodes a member of the annexin family, a group of calcium-dependent phospholipid-binding proteins. Annexins have unique N-terminal domains and conserved C-terminal domains, which contain the calcium-dependent phospholipid-binding sites. The encoded protein is a 56-kD antigen recognized by sera from patients with various autoimmune diseases. Transcript variants encoding the same isoform have been identified.
Specifications
| Annexin A11 | |
| Polyclonal | |
| Unconjugated | |
| ANXA11 | |
| 56 kDa autoantigen; A830099O17Rik; annexin; annexin A11; Annexin A11 (Annexin XI) (Calcyclin-associated annexin 50) (CAP-50); annexin XI; Annexin-11; ANX11; Anxa11; autoantigen; autoantigen, 56-kD; calcyclin-associated annexin 50; Calcyclin-associated annexin-50; CAP50; CAP-50; OTTHUMP00000019957; OTTHUMP00000019958; RP11-369J21 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 311 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P50995 | |
| ANXA11 | |
| Recombinant protein corresponding to Human ANXA11. Recombinant protein control fragment (Product #RP-95118). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction