missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A11 Mouse anti-Human, Mouse, Rat, Rabbit, Pig, Clone: 1A3C4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68089-1-IG-20UL
This item is not returnable.
View return policy
Description
Annexin A11 Monoclonal antibody apecifically detetects Annexin A11 antigen in Human, Mouse, Rat, Rabbit, Pig Samples. It is validated for Western Blot, Immunohistochemistry, ELISA.Specifications
| Annexin A11 | |
| Monoclonal | |
| 1000 μg/mL | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| ANXA11 | |
| Annexin A11 fusion protein Ag31839 | |
| 20 μL | |
| 311 | |
| Store at -20°C. Stable for one year after shipment. | |
| IgG1 |
| Western Blot, Immunohistochemistry, ELISA | |
| 1A3C4 | |
| Unconjugated | |
| BC007564 | |
| Mouse | |
| Protein G purification | |
| Primary | |
| Human, Mouse, Rat, Rabbit, Pig | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction