missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Annexin A11 Monoclonal Antibody (24GB6225)

Mouse Monoclonal Antibody
£172.00 - £488.00
Specifications
| Antigen | Annexin A11 |
|---|---|
| Clone | 24GB6225 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30276191
![]() |
Invitrogen™
MA557335 |
100 μL |
£488.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30276623
|
Invitrogen™
MA557336 |
20 μL |
£172.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Annexin A11 Monoclonal Antibody for Western Blot
This gene encodes a member of the annexin family, a group of calcium-dependent phospholipid-binding proteins. Annexins have unique N-terminal domains and conserved C-terminal domains, which contain the calcium-dependent phospholipid-binding sites. The encoded protein is a 56-kD antigen recognized by sera from patients with various autoimmune diseases. Transcript variants encoding the same isoform have been identified.Specifications
| Annexin A11 | |
| -20°C | |
| Monoclonal | |
| Liquid | |
| IgG1 | |
| Human, Mouse, Rat | |
| ANXA11 | |
| 56 kDa autoantigen; A830099O17Rik; annexin; annexin A11; Annexin A11 (Annexin XI) (Calcyclin-associated annexin 50) (CAP-50); annexin XI; Annexin-11; ANX11; Anxa11; autoantigen; autoantigen, 56-kD; calcyclin-associated annexin 50; Calcyclin-associated annexin-50; CAP50; CAP-50; OTTHUMP00000019957; OTTHUMP00000019958; RP11-369J21 | |
| ANXA11 | |
| Primary | |
| Affinity chromatography |
| 24GB6225 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P50995, P97384 | |
| 11744, 290527, 311 | |
| Recombinant protein of human ANXA11 (aa 1-505). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title