missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB100-78588
This item is not returnable.
View return policy
Description
Annexin A11 Polyclonal specifically detects Annexin A11 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Annexin A11 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| ANXA11 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human Annexin XI. The exact sequence is proprietary. | |
| Protein A purified | |
| RUO | |
| Primary | |
| ANXA11. Human. Predicted cross-reactivity based on sequence identity: Japanese Medaka (85%). | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 3.95mg/mL | |
| Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence | |
| annexin A11, Annexin-11, ANX1156-kD, CAP50, CAP-50 | |
| Rabbit | |
| 54 kDa | |
| 0.1 mL | |
| Cancer | |
| 311 | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction