Learn More
Invitrogen™ Annexin A10 Monoclonal Antibody (K94033_12H4), Invitrogen™
Mouse Monoclonal Antibody
£380.00 - £570.00
Specifications
| Antigen | Annexin A10 |
|---|---|
| Clone | K94033_12H4 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30207488
|
Invitrogen™
MA556082 |
200 μL |
£570.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30206340
|
Invitrogen™
MA555702 |
50 μL |
£380.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: SMYGRDLIGD MREQLSDHFK DVMAGLMYPP PLYDAHELWH AMKGVGTDEN CLIEILASRT NGEIFQMREA YCLQYSNNLQ EDIYSETSGH FRDTLMNLVQ GTREEGYTDP AMAAQDAMVL WEACQQKTGE HKTMLQMILC NKSYQQLRLV FQEFQNISGQ DMVDAINECY DGYFQELLVA IVLCVRDKPA YFAYRLYSAI HDFGFHNKTV IRILIARSEI DLLTIRKRYK ERYGKSLFHD IRNFASGHYK
Annexin A10 Monoclonal antibody specifically detects Annexin A10 in Human samples. It is validated for Western BlotSpecifications
| Annexin A10 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| annexin 14; annexin A10; Annexin-10; annexin-14; ANX14; Anxa10; RGD1560956 | |
| Recombinant human ANXA10 (immunogen range is 61-310). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| ANXA10 |
| K94033_12H4 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9UJ72 | |
| 11199 | |
| IgG1 | |
| Protein A | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.